Nalgene® InVitro™ Biotainer Bottles, Sterile, PETG, Thermo Scientific
Supplier: Thermo Fisher Scientific
3025-42 3005-42 3005-70 3110-42 3110-35 3230-42 3230-20
82028-478CS 819.07 CAD
82028-478 82028-480 82028-482 82028-484 82028-486 82028-488 82028-490
Nalgene® InVitro™ Biotainer Bottles, Sterile, PETG, Thermo Scientific
Bottles Plastic Bottles
Nalgene® InVitro™ Biotainer Bottles are designed specifically for bio-pharmaceutical manufacturers.
The bottles are made from crystal clear PETG, feature a space-saving square shape with ribbed hand grips (except 125 ml), and the closures feature a polyethylene foam core liner.
They reduce the risk of carry-over contamination by eliminating the cost of cleaning, sterilizing and associated validation.
Certifications: Meet USP 87, 88. Lot tested for pyrogenicity.