Nalgene® Powder Funnels, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
4252-0065 4252-0080 4252-0100 4252-0150
30255-022CS 348.26 CAD
30255-022 30255-044 30255-066 30255-088
Nalgene® Powder Funnels, Polypropylene, Thermo Scientific
Funnels Powder Funnels
Nalgene® Polypropylene Powder Funnels are used for transferring powders
The parallel stem minimizes the bridging of powder while the external ribbing prevents air locks. They are chemical resistant to a wide range of lab chemicals and reagents. These funnels are a shatter-proof alternative to glass.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)