Nalgene® PFA Beakers with Handles, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS1511-1000 DS1511-3000
13917-574EA 1045.96 CAD
13917-574 13917-578
Nalgene® PFA Beakers with Handles, Thermo Scientific
Beakers
Nalgene® PFA Beakers with Handles feature extraordinary chemical resistance and can be used repeatedly for high-purity applications
  • Rigid and translucent, enabling easy and consistent viewing of liquid levels
  • Molded in graduations in both oz. and mL that won't wear off with continuous use or washings
  • PFA is "non-stick" making the beaker easy to clean and resistant to contamination
The beakers can be used (and re-used) with strong oxidizing agents, acids, hydrocarbons and ketones. They can withstand temperatures from -270°C to +250°C, but are not forMore Product Information