Gastight® 1700 Series Waters WISP Autosampler Syringes, Hamilton Company
Supplier: Hamilton
80024 80020
60376-265EA 427.05 CAD
60376-265 72310-324
Gastight® 1700 Series Waters WISP Autosampler Syringes, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
This line of syringes is designed to work with Waters WISP HPLC autosamplers. These syringes have a Cheminert (CX) termination and are designed to fit directly into the instrumentation removing the need for a needle.
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
Syringe Termination:
Cheminert (CX) – This termination is a 1/4-28 fitting designed to fit directly into instrumentation. The X-style, meaning plunger stop, on this syringe is designed to limit wear and tear on the plunger tip.
Each is manufactured to achieve the highest level of accuracy and precision possible.