Disposal Bags, Polypropylene, Greiner Bio-One
Supplier: Greiner Bio-One
643201 646201 646401 649401 649201
82050-602CS 340.09 CAD
82050-602 82050-610 82050-612 82050-616 10859-854
Disposal Bags, Polypropylene, Greiner Bio-One
Bags Waste Disposal Bags
Greiner Bio -One disposal bags are ideal for the disposal of contaminated laboratory items.
  • Autoclavable
  • Polypropylene construction
  • Disposal bags may be marked with 'biohazard' upon request
Standard bags are made of polypropylene foil for sterilization in the steam autoclave.
Also available upon request with imprint 'biohazard'.
Oxygen transport rate of 50 µm PP foil.