PCR Plate Sealers, Greiner Bio-One
Supplier: Greiner Bio-One
676070 676090 676001 676040
82050-994CS 759.34 CAD
82050-994 82050-998 82050-992 82051-632
PCR Plate Sealers, Greiner Bio-One
Seals Microplate Seals
Greiner Bio-One offers four different varieties of adhesive microplate sealing films and foils.
  • Acrylate adhesive coating
  • Transparency differs between models
  • Different seals are ideal for different applications
EASYseal™ is a transparent sealer used for protection against contamination and evaporation
All plate sealers are coated with an acrylate adhesive. VIEWseal™ is coated with a pressure sensitive silicone adhesive.
SILVERseal™ is an aluminum sealer ideal for PCR applications and storage of sample material and active agents. VIEWseal™ is highly transparent and is most suited for protein crystallography. AMPLIseal™ is ideal for real-time PCR.