Nalgene® Wide-Mouth Square Bottles, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
312114-0002 312114-0006 312114-0008 312114-0016 312114-0032
16059-902CS 2348.51 CAD
16059-902 16059-904 16059-906 16059-908 16059-910
Nalgene® Wide-Mouth Square Bottles, HDPE, Bulk Pack, Thermo Scientific
Bottles Roller Bottles
Nalgene® wide-mouth high density polyethylene square bottles are leak proof and have a square shape that saves shelf space.
The bottles have uniform walls that are resistant to splitting or puncturing, and have a wide mouth for easy filling and pouring.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.