Envoy® Overnight Ready Engineered Shippers, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
ERSS1SEU ERWS1SEU ERSS1LEU ERWS1LEU ERSS2SEU ERWS2SEU ERSS2LEU ERWS2LEU
89205-738EA 90.46 CAD
89205-738 89205-740 89205-742 89205-744 89205-746 89205-748 89205-750 89205-752
Envoy® Overnight Ready Engineered Shippers, Sonoco ThermoSafe
Mailers and Shippers
A simple, economical solution for less complex shipping needs.
  • Up to 54 hours of protection
  • Ideal for one to two day deliveries
  • Easy to assemble
1-Day Shippers offer 30 hours of protection
Perfect for last mile pharmacy shipments. Ideal for one- to two-day delivery; this series hold product between 2 and 8°C (36 and 46°F). Seasonal packouts are simple to follow and easy to assemble. Reusable gels are environmentally friendly.
2-Day Shippers protect for 54 hours.
Certifications: Compatible to ISTA-5.
Order Now
SPECIFICATIONS
Environmentally Preferable