VITLAB® Powder Funnels, Polypropylene, BrandTech
Supplier: Brandtech
70794 70894 70994 71094 71194
83009-024PK 103.28 CAD
83009-024 83009-025 83009-026 83009-027 83009-028
VITLAB® Powder Funnels, Polypropylene, BrandTech
Funnels Powder Funnels
Translucent polypropylene funnels have a wider diameter stem for easy pouring of granulates.
  • Wide diameter stem for easy transfer of granulates
  • Autoclavable at 121 °C (250 °F)
Maximum operating temperature: 125 °C (257 °F).