FOCUS™ FASTsilver™ Electrophoresis Gel Staining Kit, G-Biosciences
Supplier: G-Biosciences
G-Biosciences' FOCUS™ FASTsilver™ produces crystal clear backgrounds and maximal peptide recovery needed for critical analysis by mass spectrometry
However, in traditional silver staining kits, silver ions are present, which inhibit proteolytic digestion. G-Biosciences' FOCUS™ FASTsilver™ is supplied with a highly efficient silver ion removal reagent called SilverOUT™. G-Biosciences' SilverOUT™ completely removes the silver ions that permit peptide digestion and extraction for maximal recovery.
G-Biosciences' FOCUS™ FASTsilver™ produces high quality silver staining without the use of glutaraldehyde, a common sensitizer in silver stains that prevents complete digestion and recovery. This results in no lysine modification, protein cross linking or reduced peptide recovery. FOCUS™ FASTsilver™ allows enhanced protease digestion and efficient recovery of digested peptides.
FOCUS™ FASTsilver™ detects proteins in less than 90 minutes and produces maximum sensitivity (~1 ng Protein or ~0.3 ng Nucleic Acid) results. For mass spectrometry analysis, complete proteolytic digestion and recovery of peptides is required for optimal analysis.