LabSafe™ GEL Blue for Staining of Polyacrylamide Gels, G-Biosciences
Supplier: G-Biosciences
LabSafe GEL Blue™ is an enhanced protein stain based on Coomassie dye that offers unsurpassed sensitivity and rapid band visualisation.
- Sensitive, 4-8ng BSA
- Saves time, band develops within 3 minutes
- Avoids the use of methanol, acetic acid, or other toxic agents
- Works with native PAGE, SDS-PAGE, isoelectric focusing, or 2D gels
LabSafe GEL Blue™ is an ultra sensitive stain that is able to detect as little as 4ng protein. Protein bands are visible in as little as 3 minutes, with maximal staining achieved in an hour. The figure shows that LabSafe GEL Blue™ can detect protein levels as low as 4ng BSA. 60 to 1000ng BSA is detectable in 5 to 10 minutes, and the low levels of BSA (<8ng) become clearly visible when washed in water.
Works with native PAGE, SDS-PAGE, denaturing, isoelectric focusing, or 2D gel. Mass spectrometry compatible.