Advanced TC™ Treated Cell Culture Flask, Standard Cap, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
658970 660960
89187-994CS 1275.26 CAD
89187-994 89204-778 89501-638 89501-640
Advanced TC™ Treated Cell Culture Flask, Standard Cap, Sterile, Greiner Bio-One
Flasks Cell Culture Flasks
Advanced Tissue Culture (TC™) treated Cell Culture Flasks, made from virgin polystyrene, allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
  • Virgin polystyrene construction
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Sterile
  • Blue filter screw cap
Advanced TC™ surface treatments provide the conditions for improved cell adherence, accelerated cell expansion and create an increase in overall cell yield.
Advanced TC™ Standard Cell Culture Flasks are available in four different sizes with a standard blue screw cap.
Sterilized by radiation, DNase- Free, RNase- Free, human DNA - Free, nonpyrogenic and noncytotoxic.