Destaining solution, Blue-OUT™ for mass spectrometry
Supplier: G-Biosciences
|
Danger
|
786-683
786-683
CA89167-894EA
374.92
CAD
CA89167-894
89167-894
Destaining solution, Blue-OUT™ for mass spectrometry
Destaining solution
Blue-OUT™ for the removal of Coomassie and fluorescent stain prior to mass spectrometry
Blue-OUT™ is a unique product that washes the interfering Coomassie and fluorescent protein stains from gel pieces prior to their treatment for protein sequencing or mass spectrometry.
Blue-OUT™ is also supplied in the InGel™ Blue kit for the tryptic digestion and peptide extraction of Coomassie stained proteins.
Storage Temperature:
Ambient |
UN:
1648 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...