BLOK™ Casein, 1% Casein Protein Blocking Solution, G-Biosciences
Supplier: G-Biosciences
786-194 786-196
CA82022-634EA 735.21 CAD
CA82022-634 CA82022-638
BLOK™ Casein, 1% Casein Protein Blocking Solution, G-Biosciences
Buffers Electrophoresis Buffers Blocking Buffers
G-Biosciences' BLOK™ Casein is a purified protein, used for blocking free binding sites in western blot, ELISA plate well, immunohistochemistry and other immunochemical applications.
G-Biosciences' BLOK™ Casein is supplied in a ready-to-use 1% (w/v) casein protein blocking solution. It is available in your choice of TBS or PBS buffers.
Ordering information: Accessories available.