Rodent Containment, Plas-Labs™
Supplier: Plas-Labs
900-RCS/EXP 900-RCL/EXP
CA10718-040EA 6810.67 CAD
CA10718-040 CA10718-042
Rodent Containment, Plas-Labs™
Animal Supplies Animal Cages
Our chamber may be used as an open or closed system
If an open system (free flow of air) is desired, align the holes on the top cone with the main clear cylinder. If not, misalign the holes to affect the closed system. Once misaligned, simply tape the top and the bottom to the main cylinder.
Order Now
SPECIFICATIONS
Environmentally Preferable