FISH-Blocker™ Non-Animal Sera Protein Blocking Agent, G-Biosciences
Supplier: G-Biosciences
786-674 786-675
CA89167-880EA 976.3 CAD
CA89167-880 CA89167-882
FISH-Blocker™ Non-Animal Sera Protein Blocking Agent, G-Biosciences
Buffers Electrophoresis Buffers Blocking Buffers
G-Biosciences' FISH-Blocker™ is uses a fish protein as the primary blocking agent to eliminate cross reactivity
G-Biosciences' FISH-Blocker™ is one of the best blocking agents for immunoassays. FISH-Blocker™ offers an alternative to milk-based blocking agents, minimizing the risk of non-specific binding of antibodies during the immunodetection process, and lowering the background.
G-Biosciences' FISH-Blocker™ is ready-to-use, and suitable for Western blot and ELISA applications. FISH-Blocker™ is offered either in TBS (Tris-buffered saline) or in PBS (phosphate-buffered saline) at pH 7.5.
Using a non-mammalian protein eliminates or minimizes the interaction of antibodies raised in mammals.
Ordering information: Accessories available.