OptiBlaze™ ELISA Chemiluminescent Detection System, G-Biosciences
Supplier: G-Biosciences
786-302 786-539
CA82023-272EA 1980.94 CAD
CA82023-272 CA95043-540
OptiBlaze™ ELISA Chemiluminescent Detection System, G-Biosciences
Assays ELISAs
G-Biosciences' OptiBlaze™ ELISA femto-HRP and femto-AP are stabilized ultra sensitive luminol and 1,2 dioxetane based horseradish peroxidase or alkaline phosphatase substrate for the detection of HRP or AP-conjugated antibodies during ELISA.
G-Biosciences' OptiBlaze™ ELISA femto-HRP or OptiBlaze™ ELISA femto-AP are chemiluminescent detection systems for Enzyme-Linked Immunosorbent Assay (ELISA). The chemiluminescent substrates provided in the kit are ultra sensitive substrates developed for luminometer-based applications, specific for horseradish peroxidase or alkaline phosphatase labeled antibodies.
G-Biosciences' OptiBlaze™ ELISA kit components are enough for performing 1,000 reactions as per the protocol.
Ordering information: Accessories available.