Well-Coated™ Nickel for Protein/Peptide Binding, G-Biosciences
Supplier: G-Biosciences
786-749 786-768 786-769
CA89167-970EA 1027.7 CAD
CA89167-970 CA89168-008 CA89168-010
Well-Coated™ Nickel for Protein/Peptide Binding, G-Biosciences
Microplates HTS Microplates
G-Biosciences' Well-Coated™ Nickel plates are designed to specifically bind 6X histidine (polyhistidine) tagged proteins and peptides.
G-Biosciences' Well-Coated™ Nickel plates can be used for the isolation of polyhistidine-tagged proteins direct from bacterial lysates and then used for subsequent ELISA protocols.
The wells are coated to a 200µL depth and are supplied pre-blocked in our proprietary Superior™ Blocking Buffer. The clear, white and black plates are offered for colorimetric, chemiluminescence and fluorescent detection systems, respectively.
Ordering information: EachMore Product Information