OPTI-SOLV® Mini Filters
Supplier: Optimize
10-04-00095 10-04-00097 10-04-02768
77100-052PK 1175.33 CAD
77100-052 77100-054 89209-702
OPTI-SOLV® Mini Filters
Filters Precolumn Filters and Frits
The OPTI-SOLV® Mini Filter provides low-impact filtration in a package no larger than a finger-tight fitting.
  • Hand-tight - no tools required 
  • Rated to 6,000psi
  • Designed to filter out particles to extend column life 
  • Perfect Zero-Dead-Volume connections every time 
  • Floating stem automatically adjusts to any depth port 
Use the Mini Filter to extend the life of analytical columns or before mass spectrometers as a last line of defense against debris. Perfect zero-dead-volume connections are obtained every time with the patentedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable