OPTI-SOLV® OPTI-SOLV® In-Line Kits
Supplier: Optimize
10-04-00383 10-04-00386 10-04-00390 10-04-00391 10-04-02371 10-04-02372
77100-066EA 1341.12 CAD
77100-066 77100-068 77100-070 77100-072 89521-908 89521-910
OPTI-SOLV® OPTI-SOLV® In-Line Kits
Filters In-Line Filters
The user-friendly hand-tight and wrench-tight OPTI-SOLV® In-Line filter system are well suited for a number of applications, such as high temperature, high-pressure environments where quick changes are imperative.
  • Rated to 6,000 psi
  • 0.2 µl total swept volume
  • Versatile for any in-line filtration application
  • Prevents damage to downstream components
  • Incorporates Free-Turn® architecture (*Hand-Tight Version)
  • Plumb once - change filter elements without disconnecting from system
The unique swivel design allows frits to be changed without removing theMore Product Information