2-Propanol ≥99.0% USP, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH2032-1GLP BDH2032-200LP
BDH2032-1GLPCS 551.82 CAD
BDH2032-1GLP BDH2032-200LP
2-Propanol ≥99.0% USP, VWR Chemicals BDH®
2-Propanol
Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (% vol) 99.0% Min.
Water (wt. %) 0.5% Max
Specific Gravity at 25º/25°C 0.783 – 0.787
Refractive Index at 20ºC 1.376 – 1.378
Description To Pass
Solubility To Pass
Identification by IR: FTIR To Pass
Identification by IR: Retention Time, GC To Pass
Nonvolatile Residue 2.5 mg (0.005%) Max.
Acidity (mL of 0.02 NaOH) 0.7 mL
Color, Pt-Co 10 Max.
Residual Solvents To Report
Limit of Volatile Impurities : Individual Impurities, % 0.10 Max.
Limit of Volatile Impurities : Total Impurities, % 1.0 Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR