OPTI-SOLV® Analytical Solvent Reservoir Filters
Supplier: Avantor
77100-036EA 267.43 CAD
77100-036 77100-038 77100-040 77100-060 77100-062 77100-064 89209-686 89521-906
OPTI-SOLV® Analytical Solvent Reservoir Filters
Filters Liquid Process Filters
A solid PTFE body keeps OPTI-SOLV® filter at the base of the bottle where a bottom-mounted titanium frit provides unrestricted solvent access.
  • Solid PTFE body keeps OPTI-SOLV filter at the base of the bottle
  • Bottom-mounted titanium frit provides unrestricted solvent access
  • Smooth flow path and conical design does not trap air
  • No tools or fittings required
As always, optimize makes installation as simple as possible no tools or fittings required. OPTI-SOLV reservoir filters employ a unique conical design inside the filter housing to prevent air bubbles from getting trapped and disturbing your analysis. While the availability of HPLC-grade solvents diminishes the need for pre-filtering, it is still imperative that a filtration device be installed in the reservoir. Placing a filter in the reservoir is an economical way to filter particles that may result from buffer salt precipitation, airborne dust, improperly cleaned glassware, or microbial contamination; none of which are remedied by the use of HPLC-grade solvents.
Delivery information: Includes five feet of pre-installed PTFE tubing.