Well-Coated™ Sulfhydryl Binding for Protein/Peptide Binding, G-Biosciences
Supplier: G-Biosciences
786-755 786-780 786-781
CA89167-982EA 681.64 CAD
CA89167-982 CA89168-032 CA89168-034
Well-Coated™ Sulfhydryl Binding for Protein/Peptide Binding, G-Biosciences
Microplates HTS Microplates
G-Biosciences' Well-Coated™ Sulfhydryl Binding plates are designed to specifically bind free sulfhydryls of peptides, proteins and other molecules
G-Biosciences' Well-Coated™ Sulfhydryl Binding plates are maleimide activated plates that react with free sulfhydryls to form stable thioether bonds at pH 6.5-7.5. pH >7.5 significantly increases the reaction of amines with the maleimide groups.
The wells are coated to a 100µL depth and are supplied pre-blocked in our proprietary Superior™ Blocking Buffer. The clear, white and black plates are offered for colorimetric, chemiluminescenceMore Product Information