Well-Coated™ Antibody (Rabbit), G-Biosciences
Supplier: G-Biosciences
786-741 786-760 786-761
CA89167-954EA 893.54 CAD
CA89167-954 CA89167-992 CA89167-994
Well-Coated™ Antibody (Rabbit), G-Biosciences
Microplates HTS Microplates
G-Biosciences' Well-Coated™ Antibody plates are designed to specifically bind either mouse or rabbit IgG making them suitable for binding assays using low quantities of antibodies or antibodies that denature on direct binding to polystyrene plates.
An added advantage of using G-Biosciences' Well-Coated™ Antibody plates is that the specificity to IgG means purified antibodies are not essential.
G-Biosciences' Well-Coated™ Antibody plates are suitable for direct, indirect, competitive and sandwich assays. The wells are coated to a 100µL depth and are supplied pre-blocked in ourMore Product Information