Well-Coated™ Antibody (Mouse), G-Biosciences
Supplier: G-Biosciences
786-739 786-758 786-759
CA89167-950EA 893.27 CAD
CA89167-950 CA89167-988 CA89167-990
Well-Coated™ Antibody (Mouse), G-Biosciences
Microplates HTS Microplates
G-Biosciences' Well-Coated™ Antibody plates are designed to specifically bind either mouse or rabbit IgG making them suitable for binding assays using low quantities of antibodies or antibodies that denature on direct binding to polystyrene plates.
An added advantage of using G-Biosciences' Well-Coated™ Antibody plates is that the specificity to IgG means purified antibodies are not essential.
G-Biosciences' Well-Coated™ Antibody plates are suitable for direct, indirect, competitive and sandwich assays. The wells are coated to a 100µL depth and are supplied pre-blocked in our proprietary Superior™ Blocking Buffer. The clear, white and black plates are offered for colorimetric, chemiluminescence and fluorescent detection systems, respectively.
Ordering information: Accessories available.