Nalgene® Square Media Bottles with Closures, PETG, Nonsterile Tray Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
322020-0030 322020-0060 322020-0125 322020-0250 322020-0500 322020-1000 322020-2000
16059-734CS 905.48 CAD
16059-734 16059-736 16059-738 16059-740 16059-742 16059-744 16059-746
Nalgene® Square Media Bottles with Closures, PETG, Nonsterile Tray Pack, Thermo Scientific
Bottles Cell Culture Bottles
Nalgene® polyethylene terephthalate copolyester square media bottles are specifically designed for packaging and shipping liquid media, buffers, and sera.
  • Non sterile
  • Leak proof
  • Molded-in graduations
These bottles are heavy-walled, durable, shatter-resistant, and provide excellent gas barrier properties.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: Bulk packed for packaging applications.