Nalgene® Square Media Bottles with Closure, Amber PETG, Non-Sterile Tray Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
322021-0030 322021-0060 322021-0125
16059-748CS 1061.06 CAD
16059-748 16059-750 16059-752
Nalgene® Square Media Bottles with Closure, Amber PETG, Non-Sterile Tray Pack, Thermo Scientific
Bottles Cell Culture Bottles
Nalgene® amber polyethylene terephthalate copolyester square media bottles are non sterile, nonpyrogenic, noncytotoxic, and have molded-in graduations.
  • Heavy-walled
  • Shatter resistant
  • Durable
They have a square design which helps save space.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: Bulk packed for packaging applications.