Nalgene Wide-Mouth Packaging Bottles, PPCO, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
312187-0001 312187-0002 312187-0004 312187-0008 312187-0016 312187-0032
16059-584CS 1822.21 CAD
16059-584 16059-586 16059-588 16059-590 16059-592 16059-594
Nalgene Wide-Mouth Packaging Bottles, PPCO, Bulk Pack, Thermo Scientific
Bottles Roller Bottles
Nalgene® wide-mouth polypropylene copolymer packaging bottles are leak proof and have wide-mouths which allows for easy filling and dispensing of powders.
These bottles come with polypropylene closures
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)