Nalgene® Rectangular Bottles, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
312007-0004 312007-0008 312007-0016 312007-0032 312009-0004 312009-0008 312009-0032
16059-818CS 1352.48 CAD
16059-818 16059-820 16059-822 16059-824 16059-826 16059-828 16059-832
Nalgene® Rectangular Bottles, HDPE, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® high density polyethylene rectangular bottles come with polypropylene closures.
Bottles are available in both translucent and amber. Translucence allows easy viewing of liquid levels. Amber reduces UV light transmissions to protect light-sensitive liquids.
All the bottles are leak proof, and feature a space-saving rectangular design. These bottles feature uniform walls which are resistant to splitting or puncturing.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information:More Product Information