Pasteur and Serum Pipettes, Greiner Bio-One
Supplier: Greiner Bio-One
612301 700361 700370 612362
82050-498CS 333.04 CAD
82050-498 82051-088 82051-090 89094-392 89094-394
Pasteur and Serum Pipettes, Greiner Bio-One
Pipettes Pasteur/Transfer Pipettes
Greiner Bio-One Pasteur and Serum Pipettes are ideally suited for the rapid transfer of liquids.
  • Sterile or nonsterile
  • Provided with or without graduations
  • Available in a range of maximum fill volumes
Pipettes are available sterile or nonsterile.
Product numbers 700370 and 700361 are designed without graduations.