Contrad 100, Low-Foaming Liquid Detergent, Decon Labs
Supplier: Decon Labs
1503 1504 1506
CA89166-412EA 1175 CAD
CA89166-412 CA89233-158 CA89233-160
Contrad 100, Low-Foaming Liquid Detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Contrad 100 is a low-foaming alkaline liquid detergent formulated for Clean-In-Place systems
Effective in removing a wide variety of contaminants from stainless steel, glass, and plastic surfaces. Its low-foaming characteristics prevent pump cavitation and excessive residual foam. Contrad 100 is free-rinsing, phosphate-free, and disposable straight to the drain. Made with recycled packaging.