Nalgene® Wide Mouth Bottles, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
312104-0001 312104-0002 312104-0004 312104-0008 312104-0016 312104-0032 312106-0001 312106-0004 312106-0008 312106-0016 312106-0032
16059-866CS 2348.51 CAD
16059-866 16059-868 16059-870 16059-872 16059-874 16059-876 16059-890 16059-894 16059-896 16059-898 16059-900
Nalgene® Wide Mouth Bottles, HDPE, Bulk Pack, Thermo Scientific
Bottles Roller Bottles
Nalgene® high-density polyethylene bottles have excellent chemical resistance, and are guaranteed leak proof.
  • Feature a wide-mouth for ease of filling and dispensing
  • Bottles have uniform walls that resist splitting and puncturing
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: Bottles are bulk packed. Available in natural and amber. Amber bottles comply with U.S. Pharmacopoeia 23,661 requirements for maximum allowable light transmission.