BRAND Titrette® Bottletop Burette, BrandTech
Supplier: Brandtech
4760151 4760161 4760251 4760261 4760141 4760241
89094-926EA 3551.11 CAD
89094-926 89094-928 89094-930 89094-932 10111-824 10111-826
BRAND Titrette® Bottletop Burette, BrandTech
Burettes
The BRAND Titrette® is the only bottletop burette that meets Class A accuracy specifications for glass burettes
  • Meets accuracy of Class A glass burettes
  • Eliminate meniscus reading error
  • Display resolution up to 10 µl
The Titrette® bottletop burette has an innovative new measuring system which matches the Class A accuracy of glass burettes and, if needed, can offer the precision of 3 decimal places below 20mL. This is an important feature for users who need to work within the Class A error limits of glass burettes according to DIN EN ISO 385 or withinMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable