Pipette Filler, S1, Thermo Scientific
Supplier: Thermo Fisher Scientific
9501 9511 9521 9531 9541
89204-752EA 1149.95 CAD
89204-752 89204-754 89204-756 89204-758 89204-760
Pipette Filler, S1, Thermo Scientific
Pipettes Pipette Controllers
The portable S1 pipette filler is designed for use with either glass or plastic serological pipettes.
Monitor Performance – LCD display shows remaining battery life and speed indicator.
Powerful Operation – Long lasting and fast performance with Li-Ion battery and powerful motor. Recharge time is 3 hours.
Control Speed – Separate aspiration and dispensing speed setting for different applications. Number of aspiration is 8. Runtime between charges will be 15 hours and fill time is 5.5 seconds for 50 ml pipet.
Your choice of colors – Available in five colors, the S1 pipette fillerMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable