sensION+ Multi-Sensors, Hach
Supplier: Hach
LZW5059.97.0002 LZW5048.97.0002 LZW5045.97.0002
CA11028-174EA 1752.01 CAD
CA11028-174 CA11028-190 CA11028-194
sensION+ Multi-Sensors, Hach
Electrodes Multi-Parameter Electrodes
sensION+ electrodes are an all-in-one solution offering several multi-sensor style electrodes and a great fit for every application.
Convenient multi-sensors for the portable instrument provide several electrodes in one. A single probe can provide readings on a combination of pH, ORP, conductivity, and/or temperature.
The electrodes offer many features, such as ease of use, temperature re-adjustment, theoretical calibration, iso-potential adjustment or calculation and more.
Certifications: .CE.WEEE.