Radiometer Metal Electrodes, Hach
Supplier: Hach
E32M002 E31M001 E34M003 E34M003 E31M003 E32M002 B35M110 E31M001 E31M002 B35M140 B35M140
89501-052EA 802.75 CAD
89501-052 CA89501-052 89501-054 CA89501-008 89501-008 89174-320 89501-004 89174-322 CA89501-058 89501-058 CA89501-054
Radiometer Metal Electrodes, Hach
Electrodes Other Electrodes
Single and combined metal electrodes can be used for Redox measurements, Redox titrations or potentiometric techniques whereas double metal electrodes are ideal for imposed current potentiometric titration.
Radiometer Analytical’s electrode range features single, double and combined metal electrodes with platinum, silver, gold, antimony, mercury and glassy carbon sensing elements.