sensION+ Benchtop Meter Bundles, Hach
Supplier: Hach
LPV2211T.97.002 LPV2200.97.0002
CA89174-146EA 3984.48 CAD
CA89174-146 CA11028-126 CA11028-130
sensION+ Benchtop Meter Bundles, Hach
Electrochemistry Meters pH and Multi-Parameter Meters
Hach sensION+, an all-in-one system with guided menu navigation makes general electrochemistry testing fast and simple. Each system is designed to be used in a wide variety of applications and comes complete with everything you need to start testing.
A guided navigation menu allows you to follow simple prompts to set up your measurements, thus reducing the time necessary to start testing and improving the accuracy of measurements.
All benchtop kits come complete with meter, probe, integrated magnetic stirrer, electrode stand, standards, calibration flasks and all required accessories.
More Product Information