pH Electrodes for HQd Meter, Hach
Supplier: Hach
PHC10101 PHC30101 PHC10105 PHC28101
CA89174-020EA 1073.12 CAD
CA89174-020 CA89174-022 CA89174-026 CA11028-180
pH Electrodes for HQd Meter, Hach
Electrodes pH Electrodes
IntelliCAL digital probes are factory calibrated and ready to test right out of the box
The IntelliCAL PHC probe is a digital, combination pH probe with a double junction reference and built-in temperature sensor.
IntelliCAL digital probes alert the user when re-calibration is needed, but re-calibration and measurement setting re-entry is not required when moving probes between meters, making these probes ideal for a multi-user environment with multiple HQd series Laboratory and Portable meters. IntelliCAL digital probes additionally provide ultimate traceability in measurement history, storing time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number. No guesswork is needed to provide the most reliable and accurate results with each measurement.
Certifications: .CE.WEEE.