ISE Electrodes for HQd Meter, Hach
Supplier: Hach
ISENH318101 ISEF12101 ISENA38103 ISENA38101
CA11027-648EA 2798.5 CAD
CA11027-648 CA11028-152 89501-108 CA89501-108 CA89174-032
ISE Electrodes for HQd Meter, Hach
Electrodes ISE Electrodes
Hach®, the leader in water quality, has developed a customized electrochemistry solution for water quality testing that takes the guesswork out of your measurements
Factory-calibrated IntelliCAL probes are ready for use, right out of the box. Each Hach® IntelliCAL probe stores its calibration history and settings in the probe itself, allowing the probes to easily be transferred from meter to meter, minimizing probe replacement and calibration. The serial number is also stored in the probe. Probes issue alerts when re-calibration is needed, but can be moved between meters without re-calibration.
Designed for your water applications, the Hach® HQd smart probes automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time.
Ordering information: Many ISE electrodes require standards, filling solutions, ISA, etc. for electrode use. Be sure to check these requirements when ordering your Hach ISE electrode.