Benchtop HQd Meter, Hach
Supplier: Hach
HQ430D HQ411D HQ440D
CA11028-128EA 4939.78 CAD
CA11028-128 CA11028-132 CA89174-012 CA89174-060 CA89174-062 10146-096
Benchtop HQd Meter, Hach
Electrochemistry Meters pH and Multi-Parameter Meters
Designed for water quality experts, the Hach HQd benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • For testing water quality
  • Advanced laboratory meter
  • Intuitive user interface
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time.
All benchtop HQd meters provide an intuitive user interface for simple operation and accurate results, while guided calibration and check standard routines reduce calibration errors. All calibrations are stored in the IntelliCAL smart probes, and calibration history allows quick and easy change out of probes without re-calibrating. Stabilization alerts and visual measurement lock additionally ensure that you can trust in the accuracy of the results.
The ultra-bright screen and large font provide easy to read results for enhanced productivity. Internal USB ports simplify data transfer, peripherals, software updates and connection to PCs printer and keyboards.
The advanced benchtop meter is designed to withstand years of continuous lab use.
Certifications: CE.WEEE.
Ordering information: The HQ440d Benchtop Meter has two electrode connector ports. All other Benchtop HQd Meters have one electrode connector port.