Zinc acetate dihydrate 98.0-102.0%, crystals USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA
Warning

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

ZI200-12KG ZI200-100GM
CA700004-372EA 5771.99 CAD
CA700004-372 75812-646
Zinc acetate dihydrate 98.0-102.0%, crystals USP
Zinc acetate dihydrate

Zinc Acetate, Crystal, USP is used in anti-itch oitments and for treating zinc deficiencies. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 237 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256

Order Now

Specification Test Results

ASSAY [Zn(CH3COO)2.2H2O] 98.0-102.0 %
pH OF A 5% SOLUTION @ 25°C 6.0-8.0
INSOLUBLE MATTER 0.005 %
ARSENIC (As) 3 ppm
LEAD (Pb) 0.002 %
CHLORIDE (Cl) 0.005 %
SULFATES (SO4) 0.010 %
ALKALIES AND ALKALINE EARTHS 0.2 %
ELEMENTAL IMPURITIES AS REPORTED
IDENTIFICATION TO PASS TEST
RESIDUAL SOLVENTS TO PASS TEST
CERTIFIED HALAL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR