Zirconium(IV) oxide ≥99% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Zirconium dioxide, Zirconium Oxide

Z1102-125GM Z1102-12KG Z1102-2.5KG Z1102-500GM
700000-918EA 118.97 CAD
700000-918 700000-920 700000-922 700000-926
Zirconium(IV) oxide ≥99% for synthesis
Zirconium (IV) oxide

Zirconium Oxide, also known as Zirconia, is an oxide of zirconium and is used as a gemstone or diamond stimulant. It can also be used in insulation and abrasives as a refractory material. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: ZrO₂
MW: 123.22 g/mol
Boiling Pt: 4300
Melting Pt: 2680
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR