Water ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Hydrogen oxide

W1000-1LT W1000-20LTCU W1000-4LTPL
CA700002-162EA 841 CAD
CA700002-162 CA700002-166 CA700002-168
Water ACS for synthesis
Water

Water, Reagent, ACS is used in many chemical reactions. Due to its ability to dissolve many ionic compounds, water is a common solvent in inorganic reactions. It is, however, rarely used as a reaction solvent since it is amphoteric and is not able to dissolve reactants very well. As an ACS grade Reagent, Spectrum Chemical manufactured water is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1.000
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR