Vanadyl sulfate hydrate ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Vanadium oxide sulfate hydrateBasic vanadium (IV) sulfate hydrateVanadium(IV) oxide sulfate hydrateVanadium (IV) sulfate oxide hydrateVanadium(IV) oxide sulphate hydrateVanadium (IV) sulphate oxide hydrateBasic vanadium (IV) sulphate hydrateVanadium oxide sulphate hydrate

V1020-100GM V1020-100G V1020-25GM V1020-500GM
CA11027-362EA 730.7 CAD
700001-576 CA11027-362 700001-570 700001-572
Vanadyl sulfate hydrate ≥97%
Vanadyl sulphate hydrate

Vanadyl Sulfate, Hydrate is an inorganic compound of the trace mineral Vanadium. These materials may or may not have a Certificate of Analysis available.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: O₅SV·H₂O
MW: 163.00 g/mol (anhy)
Density: 3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149933
CAS Number: 123334-20-3
UN: 2931
ADR: 6.1,II
Merck Index: 12,10063

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR