Triethanolamine (trolamine) ≥99% NF

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: 2,2',2''-NitrilotriethanolTris-(2-hydroxyethyl)amine

TR143-500CS6 TR143-200LTB TR143-4LTGL TR143-20LT TR143-20LT TR143-4CS4 TR143-500MLG
CA700006-962EA 5481.37 CAD
700006-962 95036-484 700006-960 CA700006-962 95036-488 95036-486 95036-490
Triethanolamine (trolamine) ≥99% NF
Triethanolamine (Trolamine)

Trolamine, NF is used as an emulsifying excipient in hand and body lotions, shaving creams, soaps, and shampoos. All Spectrum Chemical NF products, including all NF Trolamine products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 335.4 °C (1013 hPa)
Melting Pt: 21.6 °C
Density: 1.13
Flash Pt: 179 °C (closed cup)
MDL Number: MFCD00002855
CAS Number: 102-71-6
Merck Index: 13,09739
https://ca.vwr.com/stibo/web/std.lang.all/75/12/2297512.jpg

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR