Triacetin (glycerol triacetate) ≥97% USP

Supplier: Spectrum Chemical Mfg. Corp.

TR106-20LT TR106-500MLG
CA700004-648EA 9286.01 CAD
CA700004-648 95036-384
Triacetin (glycerol triacetate) ≥97% USP
Triacetin (Glycerol triacetate)

Triacetin, USP is used as a humectant excipient. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₉H₁₄O₆
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.161
Flash Pt: 138 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008716
CAS Number: 102-76-1
EINECS: 203-051-9
Merck Index: 13,09664

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR