D-(-)-Sorbitol solution 70% in aqueous solution ≥70% USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol

SO220-20LTPL SO220-500MLG
CA89050-520EA 3003 CAD
CA89050-520 CA95036-042
D-(-)-Sorbitol solution 70% in aqueous solution ≥70% USP
D(-)-Sorbitol

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOH₂C(CH(OH))₄CH₂OH
Density: 1.280
Flash Pt: Min. 100
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR