Sodium selenite ≥95%, powder

Supplier: Spectrum Chemical Mfg. Corp.
Danger

SO206-100GM SO206-10
700002-054EA 297.83 CAD
700002-054 CA11024-426
Sodium selenite ≥95%, powder
Sodium selenite

Sodium Selenite, 99.8+ Percent, Powder is the most common selenium compound that is soluble in water. It is a solid that has no color and is a salt. Its most common use is in the glass industry but it is also used as a food supplement due to its nature as an essential element

Formula: Na₂SeO₃
Melting Pt: 710 °C
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003489
CAS Number: 10102-18-8
EINECS: 233-267-9
UN: 2630
ADR: 6.1,I
Merck Index: 12,08822

Order Now

Specification Test Results

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR