Sodium metabisulfite ≥65%, granules BP, Ph. Eur., JP, NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP CA
Danger

Synonyms: Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite

SO182-12KG SO182-45KGBL
CA700001-466EA 3772.99 CAD
CA700001-466 CA89079-014
Sodium metabisulfite ≥65%, granules BP, Ph. Eur., JP, NF suitable for use as excipient
Sodium metabisulphite

Sodium Metabisulfite, Granular, NF, EP, BP, JP.


All Spectrum Chemical NF, EP, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: Na₂S₂O₅
MW: 190.11 g/mol
Melting Pt: 150
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 65% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR