Sodium benzoate ≥99%, powder NF

Supplier: Spectrum Chemical Mfg. Corp.

SO120-12KG SO120-50KGBL
CA700005-628EA 2013.99 CAD
CA700005-628 CA89078-998
Sodium benzoate ≥99%, powder NF
Sodium benzoate

Sodium Benzoate, Powder, NF is a tablet and capsule lubricant used in the pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₆H₅COONa
Melting Pt: 410…430 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR